Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001174078.1 | NP_001167549.1 | 2 - 3 | 480 | 113 | |
NM_001174079.1 | NP_001167550.1 | 2 - 3 | 480 | 113 | ||
NM_011417.3 | NP_035547.2 | 2 - 3 | 480 | 113 | ||
GenBank mRNA | AK051530.1 | - | 2 - 3 | 269 | 113 | |
AK135308.1 | - | 2 - 3 | 376 | 113 | ||
AK142137.1 | - | 2 - 3 | 376 | 113 | ||
AK147285.1 | - | 2 - 3 | 434 | 113 | ||
AK160825.1 | - | 2 - 3 | 384 | 113 | ||
AK166837.1 | - | 2 - 3 | 385 | 113 | ||
BC014760.1 | - | 2 - 3 | 291 | 113 | ||
BC023830.1 | - | 2 - 3 | 338 | 113 | ||
BC032162.1 | - | 2 - 3 | 338 | 113 | ||
BC054056.1 | - | 2 - 3 | 322 | 113 | ||
BC060229.1 | - | 2 - 3 | 374 | 113 | ||
BC061214.1 | - | 2 - 3 | 375 | 113 | ||
BC079560.1 | - | 2 - 3 | 480 | 30533489 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |