Search
Search

Ccpg1| Gene Symbol: Ccpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114328.2 | NP_001107800.1 | 2 - 3 | 236 | 68 | |
| NM_001286544.1 | NP_001273473.1 | 2 - 3 | 236 | 68 | ||
| NM_001286545.1 | NP_001273474.1 | 2 - 3 | 236 | 68 | ||
| NM_001286546.1 | NP_001273475.1 | 2 - 3 | 236 | 68 | ||
| NM_028181.5 | NP_082457.3 | 2 - 3 | 236 | 68 | ||
| GenBank mRNA | AK007973.1 | - | 2 - 3 | 133 | 68 | |
| AK032645.1 | - | 2 - 3 | 122 | 68 | ||
| AK076325.2 | - | 2 - 3 | 163 | 68 | ||
| AK128894.1 | - | 2 - 3 | 162 | 68 | ||
| AK134675.1 | - | 2 - 3 | 133 | 68 | ||
| BC043049.1 | - | 2 - 3 | 135 | 5704185 | 68 | |
| BC082602.1 | - | 2 - 3 | 137 | 30663407 | 68 | |
| CN721780.1 | - | 3 - 4 | 349 | 68 | ||
Set Membership: |
|