Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01152449_g1
          See other SRSF1 GE Assays ›
          Gene Symbol:
          Srsf1
          Gene Name:
          serine/arginine-rich splicing factor 1
          Gene Aliases:
          1110054N12Rik, 5730507C05Rik, 6330415C05Rik, AI482334, AW491331, Asf, Sf2, Sfrs1
          Chromosome Location:
          Chr.11: 88046902 - 88053757 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01152449_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Srsf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001078167.2 NP_001071635.1 1 - 2 657 59
          NM_173374.4 NP_775550.2 1 - 2 657 59
          GenBank mRNA AK018176.1 - 1 - 2 326 59
          AK053674.1 - 1 - 2 347 59
          AK078715.1 - 1 - 2 314 59
          AK150535.1 - 1 - 2 360 59
          AK169203.1 - 1 - 2 313 59
          AK169623.1 - 1 - 2 315 59
          BC046773.1 - 1 - 2 289 5708288 59
          BC058627.1 - 1 - 2 323 6401674 59
          BY743476.1 - 1 - 2 360 59
          CF739274.1 - 1 - 2 657 59
          X66091.1 - 1 - 2 270 59

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Srsf1

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA 5'-splice site recognition
          in utero embryonic development
          regulation of transcription, DNA-templated
          mRNA processing
          regulation of translation
          transport
          RNA splicing
          regulation of mRNA stability
          regulation of mRNA splicing, via spliceosome
          mRNA transport
          cardiac muscle contraction
          nucleotide binding
          nucleic acid binding
          RNA binding
          mRNA binding
          protein binding
          poly(A) RNA binding
          RS domain binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100012785.mm.1 NM_001078167.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012785.mm.1 NM_173374.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012785.mm.1 TC1100001359.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100012787.mm.1 NM_001078167.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012787.mm.1 NM_173374.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012787.mm.1 TC1100001359.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100012788.mm.1 NM_001078167.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012788.mm.1 NM_173374.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012788.mm.1 TC1100001359.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 874198 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline