Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01157077_s1
          See other MAGEA6 GE Assays ›
          Gene Symbol:
          Magea6
          Gene Name:
          melanoma antigen, family A, 6
          Gene Aliases:
          Mage-a6
          Chromosome Location:
          Chr.X: 154924012 - 154926115 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01157077_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Magea6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020019.3 NP_064403.1 2 1096 89
          GenBank mRNA BC109339.2 - 2 1107 40052528 89
          BC116902.1 - 1 1006 40126221 89

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Magea6

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974725 NM_020019.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323456 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168331 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122703 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152822 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974725 NM_020019.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32757706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32757704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32757705 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152822 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122703 - -
          Whole Mouse Genome G4122A A_52_P551956 BC116902.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P551956 NM_020019.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P551956 BC109339.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152822 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122703 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14910668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32757705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32757706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32757705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14910668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32757704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32757706 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10607432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607456 - -
          Affymetrix HT MG-430 PM Array Plate 1422357_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10607431 - -
          Affymetrix Mouse Gene 1.1 ST Array 10607454 - -
          Affymetrix Mouse Gene 1.1 ST Array 10607443 - -
          Mouse Genome 430 2.0 Array 1422357_at - -
          Mouse Genome 430 2.0 Array 1422357_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17142_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17142_PM_at - -
          Mouse Expression Set 430 1450542_PM_s_at - -
          Mouse Expression Set 430 1450542_s_at - -
          Mouse Expression Set 430 1422357_at - -
          Mouse Expression Set 430 1422357_PM_at - -
          Mouse Genome 430A 2.0 Array 1422357_PM_at - -
          Mouse Genome 430A 2.0 Array 1422357_at - -
          Mouse Genome 430A 2.0 Array 1450542_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1450542_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1422357_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422357_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4590039 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004590039 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2687002 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2694973 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2687002 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2694973 - -
          mouse-6 v1.1 (Illumina) scl0017142.1_905-S - -
          Illumina Sentrix mouse-6.v1 bead array 4590039 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0017142.1_905-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4590039 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2694973 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2694973 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0