Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013505.3 | NP_038533.1 | 10 - 11 | 1765 | 81 | |
GenBank mRNA | AK028604.1 | - | 11 - 12 | 1805 | 81 | |
AK028636.1 | - | 10 - 11 | 1766 | 81 | ||
AK030333.1 | - | 10 - 11 | 1766 | 81 | ||
AK048547.1 | - | 10 - 11 | 1766 | 81 | ||
AK172168.1 | - | 10 - 11 | 1600 | 81 | ||
BC004663.1 | - | 4 - 5 | 652 | 81 | ||
BC057867.1 | - | 10 - 11 | 1545 | 5327748 | 81 | |
L33779.1 | - | 10 - 11 | 1564 | 81 | ||
X73885.1 | - | 3 - 4 | 483 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |