Search
Search
| Gene Symbol: Psd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289602.1 | NP_001276531.1 | 4 - 5 | 1439 | 60 | |
| NM_028707.4 | NP_082983.3 | 4 - 5 | 1436 | 60 | ||
| NR_110349.1 | 4 - 5 | 1436 | 60 | |||
| GenBank mRNA | AK034575.1 | - | 4 - 5 | 1202 | 60 | |
| AK045529.1 | - | 4 - 5 | 1164 | 60 | ||
| AK053835.1 | - | 4 - 5 | 1164 | 60 | ||
| AK149027.1 | - | 4 - 5 | 1163 | 60 | ||
| BC062930.1 | - | 3 - 4 | 1017 | 6855293 | 60 | |
| BC065053.1 | - | 4 - 5 | 1201 | 5709231 | 60 | |
| BC065055.1 | - | 4 - 5 | 1187 | 5720949 | 60 | |
| BC066026.1 | - | 4 - 5 | 1436 | 5721455 | 60 | |
| BC066036.1 | - | 4 - 5 | 1202 | 6837266 | 60 | |
Set Membership: |
|