Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01158100_m1
          See other ECSCR GE Assays ›
          Gene Symbol:
          Ecscr
          Gene Name:
          endothelial cell surface expressed chemotaxis and apoptosis regulator
          Gene Aliases:
          1110006O17Rik, ARIA
          Chromosome Location:
          Chr.18: 35713086 - 35722725 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01158100_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ecscr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033141.1 NP_001028313.1 4 - 5 413 94
          GenBank mRNA AK157485.1 - 4 - 5 413 94
          EU025067.1 - 4 - 5 349 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001180.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ecscr

          Gene Ontology Categories:

          Function(s)

          angiogenesis
          apoptotic process
          chemotaxis
          multicellular organism development
          cell differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1026777 NM_001033141.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1026777 AK003507.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1026777 AK157485.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1026777 NM_001033141.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1026777 AK003507.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1026777 AK157485.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 249183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 748014 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 953534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667096 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122550 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1026777 NM_001033141.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1026777 AK003507.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1026777 AK157485.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1026777 NM_001033141.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1026777 AK003507.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1026777 AK157485.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03449381 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08964371 - -
          Whole Mouse Genome G4122A A_52_P1026777 NM_001033141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1026777 AK003507.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1026777 AK157485.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1026777 NM_001033141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1026777 AK003507.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1026777 AK157485.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08964371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31619681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13823345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13714669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14026177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14118889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13882188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13955723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13875141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13675506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13900245 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10458305 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458308 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458311 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458310 - -
          Affymetrix HT MG-430 PM Array Plate 1437451_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10458303 - -
          Mouse Genome 430 2.0 Array 1437451_PM_at - -
          Mouse Genome 430 2.0 Array 1437451_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68545_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68545_at - -
          Affymetrix Mouse Clariom S Array PSR1800008703.mm.1 NM_001033141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008703.mm.1 NM_001033141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008703.mm.1 TC1800001180.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008702.mm.1 NM_001033141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008702.mm.1 NM_001033141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008702.mm.1 TC1800001180.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008709.mm.1 NM_001033141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008709.mm.1 NM_001033141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008709.mm.1 TC1800001180.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008704.mm.1 NM_001033141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008704.mm.1 NM_001033141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008704.mm.1 TC1800001180.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008705.mm.1 NM_001033141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008705.mm.1 NM_001033141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008705.mm.1 TC1800001180.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008708.mm.1 NM_001033141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008708.mm.1 NM_001033141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008708.mm.1 TC1800001180.mm.1 MTA TC
          Mouse Expression Set 430 1437451_at - -
          Mouse Expression Set 430 1437451_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437451_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437451_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 337868 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2650674 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2650674 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline