Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01159311_m1
          See other ANKRD7 GE Assays ›
          Gene Symbol:
          Ankrd7
          Gene Name:
          ankyrin repeat domain 7
          Gene Aliases:
          4930532L20Rik, AV278427
          Chromosome Location:
          Chr.6: 18861634 - 18879586 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01159311_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ankrd7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001167757.1 NP_001161229.1 5 - 6 945 124
          NM_029202.2 NP_083478.2 5 - 6 942 124
          GenBank mRNA AK015948.1 - 5 - 6 886 124
          AK077080.1 - 5 - 6 914 124
          BC049731.1 - 5 - 6 942 6771078 124

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ankrd7

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028305 BC049731.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028305 AK077080.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028305 NM_001167757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028305 NM_029202.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028305 AK015948.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868344 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674823 AK076359.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674823 AK165804.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674823 AK169506.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940198 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823419 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59649 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149799 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140123 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028305 BC049731.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028305 AK077080.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028305 NM_001167757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028305 NM_029202.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028305 AK015948.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24275598 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149799 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140123 - -
          Whole Mouse Genome G4122A A_51_P400279 BC049731.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400279 AK077080.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400279 NM_001167757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400279 NM_029202.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400279 AK015948.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400277 AK077080.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400277 NM_001167757.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140123 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149799 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05056490 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05056497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24275620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05726530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05056490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05794599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05050340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05050336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05794594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05056497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05056495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05794595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24275628 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10536603 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536604 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536605 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536602 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536601 - -
          Affymetrix HT MG-430 PM Array Plate 1429605_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10536600 - -
          Mouse Genome 430 2.0 Array 1429605_at - -
          Mouse Genome 430 2.0 Array 1429605_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75196_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75196_at - -
          Mouse Expression Set 430 1429605_at - -
          Mouse Expression Set 430 1429605_PM_at - -
          Murine Genome U74v2 Set 166156_PM_f_at - -
          Murine Genome U74v2 Set 166156_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1429605_at - -
          Affymetrix HT Mouse Genome 430B Array 1429605_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 610701 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2708235 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2708235 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2838382 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2838382 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0