Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01159557_s1
          See other OLFR452 GE Assays ›
          Gene Symbol:
          Olfr452
          Gene Name:
          olfactory receptor 452
          Gene Aliases:
          MOR257-5P
          Chromosome Location:
          Chr.6: 42790041 - 42790994 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01159557_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr452
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011869.1 NP_001011869.1 1 724 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr452

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137223 NM_001011869.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541569 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137223 NM_001011869.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400082 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05231775 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05231776 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05231768 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05231765 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05231774 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05231769 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05231762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05216880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05701430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05826242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05216874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05826220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05314735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05701428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05231792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05651773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05352541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05251313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05084491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05216863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400148 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537796 - -
          Affymetrix Mouse Gene 1.1 ST Array 10537795 - -
          Affymetrix Mouse Clariom S Array PSR0600003734.mm.1 TC0600000467.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600003734.mm.1 NM_001011869.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003734.mm.1 NM_001011869.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 663627 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162117 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162117 - -
          mouse-6 v1.1 (Illumina) GI_58801491-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3162117 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3162117 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline