Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01160271_s1
          See other TAS2R116 GE Assays ›
          Gene Symbol:
          Tas2r116
          Gene Name:
          taste receptor, type 2, member 116
          Gene Aliases:
          T2R16, TRB1, TRB4, Tas2r14, Tas2r16, Tas2r7, mGR16, mt2r56
          Chromosome Location:
          Chr.6: 132855438 - 132856355 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01160271_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas2r116
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053212.1 NP_444442.1 1 841 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas2r116

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          response to stimulus
          sensory perception of taste
          signal transducer activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P445320 NM_053212.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P445320 AF247734.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P445320 NM_053212.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P445320 AF247734.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139690 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P445320 NM_053212.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P445320 AF247734.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P445320 NM_053212.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P445320 AF247734.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854950 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139690 - -
          Whole Mouse Genome G4122A A_51_P445320 NM_053212.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P445320 AF247734.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P445320 NM_053212.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P445320 AF247734.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126127 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139690 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006713 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006718 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006697 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006709 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006706 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05211538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05337206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05042254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854943 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10542252 - -
          Affymetrix Mouse Gene 1.1 ST Array 10542251 - -
          Affymetrix Mouse Clariom S Array PSR0600012619.mm.1 TC0600001565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012619.mm.1 NM_053212.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012619.mm.1 NM_053212.1 Refseq
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102190435 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2436799 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2938373 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2938373 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline