Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01160274_s1
          See other TAS2R110 GE Assays ›
          Gene Symbol:
          Tas2r110
          Gene Name:
          taste receptor, type 2, member 110
          Gene Aliases:
          T2R10, Tas2r10, mt2r57
          Chromosome Location:
          Chr.6: 132868008 - 132869009 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01160274_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas2r110
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199155.2 NP_954606.2 1 234 149

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas2r110

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          signal transduction
          G-protein coupled receptor signaling pathway
          response to stimulus
          sensory perception of taste
          molecular_function
          signal transducer activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P310261 NM_199155.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P310261 NM_199155.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170325 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P310261 NM_199155.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P310261 NM_199155.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P310261 NM_199155.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P310261 NM_199155.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855030 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170325 - -
          Whole Mouse Genome G4122A A_52_P310261 NM_199155.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P310261 NM_199155.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170325 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05048593 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05048587 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05048570 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05048592 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05048564 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05048602 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05048568 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05048589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05048582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05048591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05396750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05048573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05396748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05048586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854991 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10542254 - -
          Affymetrix Mouse Gene 1.1 ST Array 10542253 - -
          Affymetrix Mouse Clariom S Array PSR0600012620.mm.1 TC0600001566.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012620.mm.1 NM_199155.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012620.mm.1 NM_199155.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 856374 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2640095 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002640095 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231231 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231231 - -
          mouse-6 v1.1 (Illumina) scl0387344.1_175-S - -
          Illumina Sentrix mouse-6.v1 bead array 2640095 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0387344.1_175-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2640095 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231231 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231231 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline