Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01160389_g1
          See other ATP5B GE Assays ›
          Gene Symbol:
          Atp5b
          Gene Name:
          ATP synthase, H+ transporting mitochondrial F1 complex, beta subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 128083307 - 128090388 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01160389_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016774.3 NP_058054.2 1 - 2 142 69
          GenBank mRNA AF030559.1 - 1 - 2 141 69
          AK010314.1 - 1 - 2 142 69
          AK083276.1 - 1 - 2 144 69
          AK084009.1 - 1 - 2 133 69
          AK145684.1 - 1 - 2 143 69
          AK148891.1 - 1 - 2 144 69
          AK150599.1 - 1 - 2 144 69
          AK151081.1 - 1 - 2 144 69
          AK151600.1 - 1 - 2 141 69
          AK152788.1 - 1 - 2 144 69
          AK152976.1 - 1 - 2 144 69
          AK153099.1 - 1 - 2 133 69
          AK159737.1 - 1 - 2 147 69
          AK159978.1 - 1 - 2 144 69
          AK160199.1 - 1 - 2 136 69
          AK160608.1 - 1 - 2 144 69
          AK164383.1 - 1 - 2 132 69
          AK166525.1 - 1 - 2 145 69
          AK166603.1 - 1 - 2 144 69
          AK166979.1 - 1 - 2 143 69
          AK167119.1 - 1 - 2 144 69
          AK167160.1 - 1 - 2 134 69
          AK167728.1 - 1 - 2 141 69
          AK167764.1 - 1 - 2 144 69
          AK168692.1 - 1 - 2 134 69
          AK168941.1 - 1 - 2 144 69
          AK169184.1 - 1 - 2 144 69
          BC013253.1 - 1 - 2 118 69
          BC037127.1 - 1 - 2 135 69
          BC046616.1 - 1 - 2 125 6494223 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001584.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp5b

          Panther Classification:

          Molecular Function -

          ATP synthase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          osteoblast differentiation
          lipid metabolic process
          ATP biosynthetic process
          transport
          ion transport
          receptor-mediated endocytosis
          negative regulation of cell adhesion involved in substrate-bound cell migration
          ATP synthesis coupled proton transport
          ATP hydrolysis coupled proton transport
          proton transport
          ATP metabolic process
          regulation of intracellular pH
          hydrogen ion transmembrane transport
          nucleotide binding
          calcium ion binding
          ATP binding
          hydrolase activity
          hydrolase activity, acting on acid anhydrides, catalyzing transmembrane movement of substances
          ATPase activity
          lipoprotein particle receptor activity
          MHC class I protein binding
          proton-transporting ATP synthase activity, rotational mechanism
          proton-transporting ATPase activity, rotational mechanism

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 913622 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline