Search
Search

Csn1s1| Gene Symbol: Csn1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286015.1 | NP_001272944.1 | 28 - 29 | 780 | 73 | |
| NM_001286016.1 | NP_001272945.1 | 29 - 30 | 804 | 73 | ||
| NM_007784.3 | NP_031810.1 | 29 - 30 | 804 | 73 | ||
| GenBank mRNA | AK085690.1 | - | 29 - 30 | 808 | 73 | |
| BC002101.1 | - | 29 - 30 | 773 | 3484624 | 73 | |
| BC003859.1 | - | 29 - 30 | 788 | 3486384 | 73 | |
| BC006024.1 | - | 28 - 29 | 764 | 3592625 | 73 | |
| BC007192.1 | - | 29 - 30 | 788 | 73 | ||
| BC008278.1 | - | 29 - 30 | 788 | 2812523 | 73 | |
| BC020321.1 | - | 10 - 11 | 258 | 73 | ||
| BC040246.1 | - | 29 - 30 | 800 | 2647887 | 73 | |
| M36780.1 | - | 28 - 29 | 743 | 73 | ||
Set Membership: |
|