Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01160618_m1
          See other KRT28 GE Assays ›
          Gene Symbol:
          Krt28
          Gene Name:
          keratin 28
          Gene Aliases:
          4733401L19Rik, Krt25d
          Chromosome Location:
          Chr.11: 99364861 - 99379930 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01160618_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krt28
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027574.1 NP_081850.1 6 - 7 1257 73
          GenBank mRNA AB288231.1 - 6 - 7 1257 73
          AK014642.1 - 6 - 7 1258 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003705.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          structural molecule activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100034191.mm.1 NM_027574.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034191.mm.1 NM_027574.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034191.mm.1 TC1100003705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034190.mm.1 NM_027574.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034190.mm.1 NM_027574.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034190.mm.1 TC1100003705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034192.mm.1 NM_027574.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034192.mm.1 NM_027574.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034192.mm.1 TC1100003705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034186.mm.1 NM_027574.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034186.mm.1 NM_027574.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034186.mm.1 TC1100003705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034188.mm.1 NM_027574.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034188.mm.1 NM_027574.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034188.mm.1 TC1100003705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034187.mm.1 NM_027574.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034187.mm.1 NM_027574.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034187.mm.1 TC1100003705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034189.mm.1 NM_027574.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034189.mm.1 NM_027574.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034189.mm.1 TC1100003705.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 298151 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0