Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01161473_s1
          See other TAS2R124 GE Assays ›
          Gene Symbol:
          Tas2r124
          Gene Name:
          taste receptor, type 2, member 124
          Gene Aliases:
          T2R24, Tas2r24, mGR24, mt2r50
          Chromosome Location:
          Chr.6: 132754730 - 132755659 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01161473_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas2r124
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207026.1 NP_996909.1 1 424 136

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas2r124

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          biological_process
          response to stimulus
          sensory perception of taste
          molecular_function
          signal transducer activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028004 NM_207026.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028004 NM_207026.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854508 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05103075 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05103078 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05103076 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05103073 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05103065 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05103059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05664699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05297086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05581939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05297088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854506 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10542244 - -
          Affymetrix Mouse Gene 1.1 ST Array 10542243 - -
          Affymetrix Mouse Clariom S Array PSR0600012609.mm.1 TC0600001561.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012609.mm.1 NM_207026.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012609.mm.1 NM_207026.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 557807 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4480300 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004480300 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2719246 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2719246 - -
          mouse-6 v1.1 (Illumina) scl0387351.1_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 4480300 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0387351.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4480300 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2719246 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2719246 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0