Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01161497_g1
          See other TAS2R109 GE Assays ›
          Gene Symbol:
          Tas2r109
          Gene Name:
          taste receptor, type 2, member 109
          Gene Aliases:
          T2R09, Tas2r9, mGR09, mt2r62
          Chromosome Location:
          Chr.6: 132980015 - 132980965 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01161497_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas2r109
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207017.1 NP_996900.1 1 313 159
          GenBank mRNA BC127604.1 - 1 310 159

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas2r109

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          biological_process
          response to stimulus
          sensory perception of taste
          molecular_function
          signal transducer activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967672 NM_207017.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967672 BC127604.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967672 NM_207017.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967672 BC127604.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855525 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06073387 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06073387 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05540836 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05310371 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05310379 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05540851 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05540854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05350534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05350532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05350543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05762853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855485 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548672 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548671 - -
          Affymetrix Mouse Clariom S Array PSR0600025838.mm.1 TC0600003290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025838.mm.1 NM_207017.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025838.mm.1 NM_207017.1 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5670112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005670112 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225944 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225944 - -
          mouse-6 v1.1 (Illumina) scl0387343.1_301-S - -
          Illumina Sentrix mouse-6.v1 bead array 5670112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0387343.1_301-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5670112 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225944 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225944 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline