Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01162710_m1
          See other SMO GE Assays ›
          Gene Symbol:
          Smo
          Gene Name:
          smoothened homolog (Drosophila)
          Gene Aliases:
          E130215L21Rik, Smoh, bnb, smoothened
          Chromosome Location:
          Chr.6: 29735497 - 29761366 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01162710_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smo
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_176996.4 NP_795970.3 8 - 9 1997 58
          GenBank mRNA AK081050.1 - 2 - 3 185 58
          BC096028.1 - 8 - 9 1516 6850065 58
          BQ947565.1 - 3 - 4 259 58
          CA749812.1 - 5 - 6 597 58
          FJ883552.1 - 8 - 9 1506 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000242.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smo

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ossification
          vasculogenesis
          osteoblast differentiation
          in utero embryonic development
          cell fate specification
          neural crest cell migration
          heart looping
          positive regulation of neuroblast proliferation
          positive regulation of mesenchymal cell proliferation
          heart morphogenesis
          determination of left/right asymmetry in lateral mesoderm
          type B pancreatic cell development
          signal transduction
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          smoothened signaling pathway
          positive regulation of hh target transcription factor activity
          multicellular organism development
          determination of left/right symmetry
          ventral midline determination
          pattern specification process
          central nervous system development
          midgut development
          positive regulation of cell proliferation
          anterior/posterior pattern specification
          dorsal/ventral pattern formation
          regulation of gene expression
          positive regulation of gene expression
          negative regulation of gene expression
          dentate gyrus development
          cerebellar cortex morphogenesis
          thalamus development
          dorsal/ventral neural tube patterning
          smoothened signaling pathway involved in ventral spinal cord patterning
          smoothened signaling pathway involved in regulation of cerebellar granule cell precursor cell proliferation
          central nervous system neuron differentiation
          cerebral cortex development
          negative regulation of epithelial cell differentiation
          hair follicle morphogenesis
          protein localization to nucleus
          multicellular organism growth
          positive regulation of protein import into nucleus
          odontogenesis of dentin-containing tooth
          negative regulation of apoptotic process
          negative regulation of DNA binding
          positive regulation of smoothened signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          astrocyte activation
          cell development
          digestive tract development
          embryonic organ development
          developmental growth
          skeletal muscle fiber development
          axon extension involved in axon guidance
          forebrain morphogenesis
          homeostasis of number of cells within a tissue
          positive regulation of epithelial cell proliferation
          protein stabilization
          myoblast migration
          negative regulation of hair follicle development
          detection of cell density by contact stimulus involved in contact inhibition
          atrial septum morphogenesis
          mammary gland epithelial cell differentiation
          epithelial-mesenchymal cell signaling
          somite development
          pancreas morphogenesis
          left/right axis specification
          mesenchymal to epithelial transition involved in metanephric renal vesicle formation
          positive regulation of branching involved in ureteric bud morphogenesis
          regulation of stem cell population maintenance
          regulation of heart morphogenesis
          signal transducer activity
          transmembrane signaling receptor activity
          G-protein coupled receptor activity
          patched binding
          drug binding
          Wnt-protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600001663.mm.1 TC0600000242.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600001663.mm.1 NM_176996.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001663.mm.1 NM_176996.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0