Search
Search

Prrt4| Gene Symbol: Prrt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101443.1 | NP_001094913.1 | 4 - 5 | 60 | ||
| GenBank mRNA | AK052319.1 | - | 4 - 5 | 60 | ||
| BC140954.1 | - | 4 - 5 | 1067 | 9055566 | 60 | |
Set Membership: |
|