Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nadsyn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ305343.1 | - | 15 - 16 | 1552 | 62 | |
AK020264.1 | - | 15 - 16 | 1552 | 62 | ||
AK041101.1 | - | 15 - 16 | 1539 | 62 | ||
AK042389.1 | - | 15 - 16 | 1512 | 62 | ||
BC018248.1 | - | 13 - 14 | 1318 | 62 | ||
BC038016.1 | - | 15 - 16 | 1498 | 3589586 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |