Search
Search

Gm8817| Gene Symbol: Gm8817 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101606.1 | NP_001095076.1 | 3 - 4 | 278 | 71 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AI592229.1 | - | 161 | Chr. - - [-] on Build | 71 | ||
Set Membership: |
|