Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01164551_g1
          See other MAZ GE Assays ›
          Gene Symbol:
          Maz
          Gene Name:
          MYC-associated zinc finger protein (purine-binding transcription factor)
          Gene Aliases:
          PUR1, Pur-1, SAF-1, SAF-2
          Chromosome Location:
          Chr.7: 127022123 - 127026553 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01164551_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Maz
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010772.1 NP_034902.1 3 - 4 1173 58
          GenBank mRNA BC004758.1 - 5 - 6 710 58
          DQ150586.1 - 2 - 3 887 58
          L04649.1 - 3 - 4 1146 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004309.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          termination of RNA polymerase II transcription
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II core promoter sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          nucleic acid binding
          DNA binding
          chromatin binding
          protein binding
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25503652 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25503657 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25503660 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25503651 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25503658 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2283207 DQ150586.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2283207 L04649.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2283207 NM_010772.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2283207 BC004758.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 AF033655.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 AK016370.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 AK045083.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 AK051283.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 AK051411.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 AK083269.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 AK083790.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 AK087398.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 AK129226.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 AK138437.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 AK139386.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 AK141535.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 AK165149.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 BC068134.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 NM_011074.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323724 U62391.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626564 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150654 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123430 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2283207 DQ150586.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2283207 L04649.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2283207 NM_010772.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2283207 BC004758.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123430 - -
          Whole Mouse Genome G4122A A_52_P673304 DQ150586.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P673304 L04649.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P673304 NM_010772.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P673304 BC004758.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P191973 DQ150586.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P191973 L04649.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P191973 NM_010772.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P191973 BC004758.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P253843 DQ150586.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P253843 NM_010772.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P253843 BC004758.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123430 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25503657 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25503660 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25503652 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25503658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25503651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25503642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06653209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05941379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25503626 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568142 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568144 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568145 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568140 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568141 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568143 - -
          Affymetrix HT MG-430 PM Array Plate 1427099_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568139 - -
          Mouse Genome 430 2.0 Array 1427099_PM_at - -
          Mouse Genome 430 2.0 Array 1427099_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17188_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17188_at - -
          Affymetrix Murine Genome 6500A Array Msa11230_at - -
          Affymetrix Mouse Clariom S Array PSR0700036155.mm.1 NM_010772.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036155.mm.1 NM_010772.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036155.mm.1 TC0700004309.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036157.mm.1 NM_010772.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036157.mm.1 NM_010772.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036157.mm.1 TC0700004309.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036154.mm.1 NM_010772.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036154.mm.1 NM_010772.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036154.mm.1 TC0700004309.mm.1 MTA TC
          Mouse Expression Set 430 1427099_PM_at - -
          Mouse Expression Set 430 1427099_at - -
          Murine Genome U74v2 Set 160355_PM_at - -
          Murine Genome U74v2 Set 160355_at - -
          Murine Genome U74v2 Set 134739_PM_r_at - -
          Murine Genome U74v2 Set 134739_r_at - -
          Mouse Genome 430A 2.0 Array 1427099_at - -
          Mouse Genome 430A 2.0 Array 1427099_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427099_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427099_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 887802 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2709839 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2709839 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2709839 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline