Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01164575_g1
          See other GM5600 GE Assays ›
          Gene Symbol:
          Gm5600
          Gene Name:
          predicted gene 5600
          Gene Aliases:
          EG434228, ENSMUSG00000073862
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01164575_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5600
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK035124.1 - 3 - 4 1933 80
          BB104418.2 - 2 - 3 279 80

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2001093 AK035124.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174440 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2001093 AK035124.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174440 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111910 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05988187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06415547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06264850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06269930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06209931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05903642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06199175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05903629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06371280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06264852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06252745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06371278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05899917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06252747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06052521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06206984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06057364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06508610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06508612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06199171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06209930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25431926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06371282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25432045 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1446893_PM_at - -
          Mouse Genome 430 2.0 Array 1446893_at - -
          Mouse Genome 430 2.0 Array 1446893_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 434228_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 434228_at - -
          Mouse Expression Set 430 1446893_PM_at - -
          Mouse Expression Set 430 1446893_at - -
          Affymetrix HT Mouse Genome 430B Array 1446893_at - -
          Affymetrix HT Mouse Genome 430B Array 1446893_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101770180 - -
          Illumina mouse-6 v1.0 expression beadchip 101770180 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243120 - -
          Illumina Sentrix mouse-6.v1 bead array 101770180 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101770180 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243120 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline