Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01164634_m1
          See other 1700001G01RIK GE Assays ›
          Gene Symbol:
          1700001G01Rik
          Gene Name:
          RIKEN cDNA 1700001G01 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 17035508 - 17137559 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01164634_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700001G01Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK005589.1 - 382 73
          AK133007.1 - 2 - 3 484 73

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044099 AK005589.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385769 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813739 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751490 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 70423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 842426 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409051 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144785 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55385 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 501534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14562 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 555955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 538857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930672 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 917312 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 612014 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245021 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 631215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 399418 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 37166 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 607435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244486 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 372880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342712 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 717961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 856773 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 318258 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 792057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683935 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852537 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722562 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 98110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 97314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144411 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126377 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27341 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45004 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64972 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044099 AK005589.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08924846 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08924878 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08924961 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08924825 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08924884 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08924978 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08924892 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08925007 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08924794 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08924920 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08924901 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100869 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114480 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08925007 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08924794 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08924846 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08924920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14073027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14073008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31526002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31526006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13716520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14081375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13944304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31526007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31526001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13805238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13942604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31526008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31526004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13625464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14108488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13625462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13784996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13784979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31525896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14108491 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0