Search
Search

Gtf3c1| Gene Symbol: Gtf3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207239.1 | NP_997122.1 | 32 - 33 | 5073 | 67 | |
| GenBank mRNA | AK147325.1 | - | 32 - 33 | 5166 | 67 | |
| AK171723.1 | - | 3 - 4 | 2135 | 67 | ||
| BC032208.1 | - | 20 - 21 | 2880 | 67 | ||
| BC067041.1 | - | 32 - 33 | 5073 | 6853285 | 67 | |
Set Membership: |
|