| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000012067.mm.1 |
NM_183264.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012067.mm.1 |
NM_183264.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012067.mm.1 |
TC1000001661.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000012071.mm.1 |
NM_183264.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012071.mm.1 |
NM_183264.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012071.mm.1 |
TC1000001661.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000012074.mm.1 |
NM_183264.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012074.mm.1 |
NM_183264.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012074.mm.1 |
TC1000001661.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000012072.mm.1 |
NM_183264.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012072.mm.1 |
NM_183264.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000012072.mm.1 |
TC1000001661.mm.1 |
MTA TC |