Search
Search

Gpr26| Gene Symbol: Gpr26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173410.3 | NP_775586.1 | 1 - 2 | 1140 | 65 | |
| GenBank mRNA | AK036100.1 | - | 1 - 2 | 1143 | 65 | |
| AK042755.1 | - | 1 - 2 | 242 | 65 | ||
| AY255585.1 | - | 1 - 2 | 324 | 65 | ||
| BC119380.1 | - | 1 - 2 | 998 | 8734129 | 65 | |
| BC120647.1 | - | 1 - 2 | 998 | 40129570 | 65 | |
Set Membership: |
|