Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01166064_g1
          See other 1700034O15RIK GE Assays ›
          Gene Symbol:
          1700034O15Rik
          Gene Name:
          RIKEN cDNA 1700034O15 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 41684346 - 41685717 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01166064_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700034O15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029671.2 NP_083947.1 1 - 2 244 101
          GenBank mRNA AK006606.1 - 1 - 2 245 101
          BC048592.1 - 1 - 2 244 6707911 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000445.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1700034O15Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286978 NM_029671.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286978 AK006606.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286978 BC048592.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286978 NM_029671.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286978 AK006606.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286978 BC048592.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 76217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 53638 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 243333 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594244 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126462 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122832 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37135 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131756 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172004 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120713 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151638 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286978 NM_029671.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286978 AK006606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286978 BC048592.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286978 NM_029671.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286978 AK006606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286978 BC048592.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24395256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24395264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24395260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24395257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24395258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24395265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24395262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24395253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24395254 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151638 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172004 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120713 - -
          Whole Mouse Genome G4122A A_51_P286978 NM_029671.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286978 AK006606.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286978 BC048592.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286978 NM_029671.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286978 AK006606.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286978 BC048592.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151638 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120713 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172004 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05576540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24395218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24395252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05080050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24395241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24395239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24395249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24395265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05576543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05576539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24395258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05576546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24395253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05576551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24395247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24395245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24395251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24395264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24395256 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537678 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537677 - -
          Affymetrix HT MG-430 PM Array Plate 1453254_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10537676 - -
          Mouse Genome 430 2.0 Array 1453254_at - -
          Mouse Genome 430 2.0 Array 1453254_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76606_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76606_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600003557.mm.1 NM_029671.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003557.mm.1 NM_029671.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003557.mm.1 TC0600000445.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600003556.mm.1 NM_029671.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003556.mm.1 NM_029671.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003556.mm.1 TC0600000445.mm.1 MTA TC
          Mouse Expression Set 430 1453254_at - -
          Mouse Expression Set 430 1453254_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453254_at - -
          Affymetrix HT Mouse Genome 430B Array 1453254_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 367945 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2613803 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2613803 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2613803 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline