Search
Search

Acsm1| Gene Symbol: Acsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054094.5 | NP_473435.1 | 3 - 4 | 479 | 79 | |
| GenBank mRNA | AB059428.1 | - | 3 - 4 | 483 | 79 | |
| AK149550.1 | - | 3 - 4 | 489 | 79 | ||
| AK149586.1 | - | 3 - 4 | 505 | 79 | ||
| BC016414.1 | - | 3 - 4 | 476 | 4238964 | 79 | |
| BC022149.1 | - | 3 - 4 | 457 | 5102652 | 79 | |
Set Membership: |
|