Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01166455_m1
          See other HIST1H2BP GE Assays ›
          Gene Symbol:
          Hist1h2bp
          Gene Name:
          histone cluster 1, H2bp
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 21787480 - 21801561 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01166455_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist1h2bp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178202.2 NP_835509.2 1 - 2 401 122
          GenBank mRNA BC061044.1 - 1 - 2 401 6742370 122
          M25487.1 - 1 - 2 362 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000179.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101132 NM_178202.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101132 AV047260.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101132 BC061044.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101132 M25487.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 507309 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101132 NM_178202.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101132 AV047260.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101132 BC061044.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101132 M25487.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874636 - -
          Whole Mouse Genome G4122A A_52_P233737 AK076671.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10910139 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10910113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10687923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10910134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10500469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11007989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11121492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10616510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11049082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28874644 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10403954 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403952 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403953 - -
          Affymetrix HT MG-430 PM Array Plate 1427762_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10403951 - -
          Mouse Genome 430 2.0 Array 1452540_a_at - -
          Mouse Genome 430 2.0 Array 1427762_x_at - -
          Mouse Genome 430 2.0 Array 1427762_PM_x_at - -
          Mouse Genome 430 2.0 Array 1452540_PM_a_at - -
          Affymetrix Mouse Clariom S Array PSR1300001187.mm.1 NM_178202.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001187.mm.1 NM_178202.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001187.mm.1 TC1300000179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001189.mm.1 NM_178202.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001189.mm.1 NM_178202.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001189.mm.1 TC1300000179.mm.1 MTA TC
          Mouse Expression Set 430 1452540_PM_a_at - -
          Mouse Expression Set 430 1452540_a_at - -
          Mouse Expression Set 430 1427762_PM_x_at - -
          Mouse Expression Set 430 1427762_x_at - -
          Murine Genome U74v2 Set 162169_r_at - -
          Murine Genome U74v2 Set 93834_at - -
          Murine Genome U74v2 Set 162169_PM_r_at - -
          Murine Genome U74v2 Set 161394_f_at - -
          Murine Genome U74v2 Set 93834_PM_at - -
          Murine Genome U74v2 Set 161394_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1452540_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1427762_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1452540_a_at - -
          Mouse Genome 430A 2.0 Array 1427762_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1427762_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1427762_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 546182 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5690079 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005690079 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1252995 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252995 - -
          mouse-6 v1.1 (Illumina) scl0319188.1_136-S - -
          Illumina Sentrix mouse-6.v1 bead array 5690079 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0319188.1_136-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5690079 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252995 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0319188.1_136-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0