Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01166846_s1
          See other LRRC4 GE Assays ›
          Gene Symbol:
          Lrrc4
          Gene Name:
          leucine rich repeat containing 4
          Gene Aliases:
          MBAG1, NGL-2, NLG-2, Nag14
          Chromosome Location:
          Chr.6: 28828126 - 28831747 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01166846_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138682.2 NP_619623.2 1 2440 141
          GenBank mRNA AK046781.1 - 1 1647 141
          DQ177325.1 - 1 2440 141

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lrrc4

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of synapse organization
          synapse organization
          postsynaptic density protein 95 clustering
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322954 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322956 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322955 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322964 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322959 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322960 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322963 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322952 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322958 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322961 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322957 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2176080 DQ177325.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2176080 AK046781.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2176080 NM_138682.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2176080 DQ177325.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2176080 AK046781.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2176080 NM_138682.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24322986 - -
          Whole Mouse Genome G4122A A_51_P337210 DQ177325.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337210 AK046781.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337210 NM_138682.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337210 AF300458.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106608 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05115623 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05758509 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05115625 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05115633 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05115620 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05758514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322963 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322956 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322960 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322964 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322958 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322959 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322957 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322955 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322961 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322952 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24322954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24323013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24323009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24323000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24323008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24323019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24323018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24323001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24323016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24323005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24323002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24323010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24322946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05766803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05875386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05875391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05532602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24323019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24323014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05782493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05419430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05495541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05532603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24323005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24323016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05875394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24323011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05333859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24323002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05787637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05766799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05766807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05824873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24323000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05846867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24323020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05766812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05115635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05203351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05660453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24322945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24323013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05115619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05766816 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10543545 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543548 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543546 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543547 - -
          Affymetrix HT MG-430 PM Array Plate 1416097_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1435832_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10543544 - -
          Mouse Genome 430 2.0 Array 1416097_PM_at - -
          Mouse Genome 430 2.0 Array 1435832_PM_at - -
          Mouse Genome 430 2.0 Array 1435832_at - -
          Mouse Genome 430 2.0 Array 1416097_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 192198_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 192198_PM_at - -
          Mouse Expression Set 430 1416097_PM_at - -
          Mouse Expression Set 430 1416097_at - -
          Mouse Expression Set 430 1435832_PM_at - -
          Mouse Expression Set 430 1435832_at - -
          Murine Genome U74v2 Set 136244_PM_at - -
          Murine Genome U74v2 Set 136244_at - -
          Mouse Genome 430A 2.0 Array 1416097_at - -
          Mouse Genome 430A 2.0 Array 1416097_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416097_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416097_at - -
          Affymetrix HT Mouse Genome 430B Array 1435832_at - -
          Affymetrix HT Mouse Genome 430B Array 1435832_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 345074 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3850041 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003850041 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246862 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246862 - -
          mouse-6 v1.1 (Illumina) scl0192198.1_50-S - -
          Illumina Sentrix mouse-6.v1 bead array 3850041 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0192198.1_50-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3850041 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2741194 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2741194 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline