Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01166868_m1
          See other 1700022A22RIK GE Assays ›
          Gene Symbol:
          1700022A22Rik
          Gene Name:
          RIKEN cDNA 1700022A22 gene
          Gene Aliases:
          4933413D07Rik
          Chromosome Location:
          Chr.15: 46324585 - 46373899 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01166868_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700022A22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045509.1 3 - 4 733 89
          GenBank mRNA AK006231.1 - 3 - 4 735 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000362.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2299976 AK016801.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320050 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 791662 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487543 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907266 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 53800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21689 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08356366 - -
          Whole Mouse Genome G4122A A_52_P426601 AK006231.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P426601 AK016801.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08356366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30188115 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432341_PM_at - -
          Mouse Genome 430 2.0 Array 1432341_at - -
          Mouse Genome 430 2.0 Array 1432341_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500002556.mm.1 NR_045509.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002556.mm.1 NR_045509.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002556.mm.1 TC1500000362.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500002555.mm.1 NR_045509.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002555.mm.1 NR_045509.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002555.mm.1 TC1500000362.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500002559.mm.1 NR_045509.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002559.mm.1 NR_045509.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002559.mm.1 TC1500000362.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500002552.mm.1 NR_045509.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002552.mm.1 NR_045509.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002552.mm.1 TC1500000362.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500002557.mm.1 NR_045509.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002557.mm.1 NR_045509.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002557.mm.1 TC1500000362.mm.1 MTA TC
          Mouse Expression Set 430 1432341_at - -
          Mouse Expression Set 430 1432341_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432341_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432341_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline