Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01167263_m1
          See other LRRC17 GE Assays ›
          Gene Symbol:
          Lrrc17
          Gene Name:
          leucine rich repeat containing 17
          Gene Aliases:
          37kDa, 4833425M04Rik, 6130400C22Rik, P37nb
          Chromosome Location:
          Chr.5: 21543527 - 21575911 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01167263_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028977.1 NP_083253.1 2 - 3 1091 75
          GenBank mRNA AK018071.1 - 2 - 3 1057 75
          BC030317.1 - 2 - 3 1010 5372009 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000173.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          osteoblast differentiation
          osteoblast proliferation
          negative regulation of osteoclast differentiation
          bone marrow development
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P301930 AK018071.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P301930 BC030317.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P301930 CD741871.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P301930 NM_028977.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P301930 AK018071.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P301930 BC030317.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P301930 CD741871.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P301930 NM_028977.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301930 AK018071.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301930 BC030317.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301930 CD741871.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301930 NM_028977.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301930 AK018071.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301930 BC030317.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301930 CD741871.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301930 NM_028977.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301930 AK018071.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301930 BC030317.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301930 CD741871.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301930 NM_028977.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301930 AK018071.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301930 BC030317.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301930 CD741871.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301930 NM_028977.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23526636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04250293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04623805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04952953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04641365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04463749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04421574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04719104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04641369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04421573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04250310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04598306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04914545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04483739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04483740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04519095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04623799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04876049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23526536 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10519999 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520002 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520000 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520001 - -
          Affymetrix HT MG-430 PM Array Plate 1454076_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429679_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10519998 - -
          Affymetrix Mouse Gene 1.1 ST Array 10528310 - -
          Mouse Genome 430 2.0 Array 1429679_at - -
          Mouse Genome 430 2.0 Array 1429679_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500001237.mm.1 TC0500000173.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500001237.mm.1 NM_028977.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001237.mm.1 NM_028977.1 Refseq
          Mouse Expression Set 430 1429679_PM_at - -
          Mouse Expression Set 430 1429679_at - -
          Affymetrix HT Mouse Genome 430B Array 1454076_at - -
          Affymetrix HT Mouse Genome 430B Array 1429679_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429679_at - -
          Affymetrix HT Mouse Genome 430B Array 1454076_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 806634 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106290138 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2511456 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline