Search
Search

Vmn1r211| Gene Symbol: Vmn1r211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134243.1 | NP_599004.1 | 1 | 402 | 123 | |
| GenBank mRNA | BC138182.1 | - | 1 | 450 | 8861201 | 123 |
| BC138183.1 | - | 1 | 450 | 8861202 | 123 | |
Set Membership: |
|