Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01167872_sH
          See other OLFR787 GE Assays ›
          Gene Symbol:
          Olfr787
          Gene Name:
          olfactory receptor 787
          Gene Aliases:
          MOR111-10
          Chromosome Location:
          Chr.10: 129461940 - 129463616 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01167872_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr787
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011822.1 NP_001011822.1 1 44 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr787

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140658 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140658 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185275 NM_001011822.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545887 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08614652 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08614642 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08614630 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08614649 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08614632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08366351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545871 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10367494 - -
          Affymetrix Mouse Gene 1.0 ST Array 10367498 - -
          Affymetrix Mouse Gene 1.1 ST Array 10367497 - -
          Affymetrix Mouse Gene 1.1 ST Array 10367493 - -
          Affymetrix Mouse Clariom S Array PSR1000012037.mm.1 NM_001011822.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012037.mm.1 NM_001011822.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012037.mm.1 TC1000001642.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 761124 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2723259 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2723259 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0