Search
Search

Fam71f2| Gene Symbol: Fam71f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101486.1 | NP_001094956.1 | 2 - 3 | 459 | 62 | |
| GenBank mRNA | AB473724.1 | - | 2 - 3 | 335 | 62 | |
| BC050974.1 | - | 2 - 3 | 459 | 62 | ||
| BC151115.1 | - | 2 - 3 | 426 | 9088028 | 62 | |
| BC151117.1 | - | 2 - 3 | 426 | 9088030 | 62 | |
Set Membership: |
|