Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01168723_m1
          See other MMP12 GE Assays ›
          Gene Symbol:
          Mmp12
          Gene Name:
          matrix metallopeptidase 12
          Gene Aliases:
          AV378681, Mmel
          Chromosome Location:
          Chr.9: 7344397 - 7360461 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01168723_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mmp12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008605.3 NP_032631.3 6 - 7 976 115
          GenBank mRNA AK089309.1 - 6 - 7 945 115
          AK089452.1 - 6 - 7 945 115
          AK089509.1 - 7 - 8 819 115
          AK089523.1 - 6 - 7 945 115
          AK090051.1 - 6 - 7 947 115
          AK150596.1 - 6 - 7 945 115
          AK151021.1 - 6 - 7 945 115
          AK151273.1 - 6 - 7 945 115
          AK151933.1 - 6 - 7 945 115
          AK155139.1 - 6 - 7 944 115
          AK155743.1 - 6 - 7 945 115
          AK155747.1 - 6 - 7 945 115
          AK155748.1 - 6 - 7 945 115
          AK155841.1 - 6 - 7 1003 115
          AK155865.1 - 6 - 7 944 115
          AK170364.1 - 6 - 7 945 115
          AK170500.1 - 7 - 8 819 115
          AK170798.1 - 6 - 7 944 115
          AK170804.1 - 6 - 7 944 115
          AK170976.1 - 6 - 7 945 115
          AK171006.1 - 6 - 7 945 115
          AK171032.1 - 6 - 7 944 115
          AK171036.1 - 6 - 7 945 115
          AK171176.1 - 6 - 7 945 115
          AK171428.1 - 6 - 7 945 115
          AK171435.1 - 5 - 6 1992 115
          AK171447.1 - 6 - 7 945 115
          AK171448.1 - 6 - 7 945 115
          AK171449.1 - 6 - 7 945 115
          AK171450.1 - 6 - 7 945 115
          AK171466.1 - 6 - 7 945 115
          AK171468.1 - 6 - 7 945 115
          AK171470.1 - 6 - 7 945 115
          AK171472.1 - 6 - 7 945 115
          AK171485.1 - 6 - 7 945 115
          AK171518.1 - 6 - 7 945 115
          AK171542.1 - 6 - 7 945 115
          AK171570.1 - 6 - 7 945 115
          BC019135.1 - 6 - 7 939 5037674 115
          CT010377.1 - 6 - 7 892 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000046.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mmp12

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          wound healing, spreading of epidermal cells
          wound healing
          positive regulation of epithelial cell proliferation involved in wound healing
          metalloendopeptidase activity
          calcium ion binding
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900000351.mm.1 NM_008605.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000351.mm.1 NM_008605.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000351.mm.1 TC0900000046.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-952lx:80/100.66.78.23:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0