Search
Search

Cabin1| Gene Symbol: Cabin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172549.3 | NP_766137.2 | 17 - 18 | 2938 | 59 | |
| GenBank mRNA | BC057551.1 | - | 17 - 18 | 2866 | 59 | |
| BC150825.1 | - | 16 - 17 | 2502 | 9087736 | 59 | |
Set Membership: |
|