Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01169300_m1
          See other MRAP2 GE Assays ›
          Gene Symbol:
          Mrap2
          Gene Name:
          melanocortin 2 receptor accessory protein 2
          Gene Aliases:
          BB633055
          Chromosome Location:
          Chr.9: 87144224 - 87184051 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01169300_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrap2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001101482.2 NP_001094952.2 2 - 3 232 119
          NM_001177731.1 NP_001171202.1 3 - 4 377 119
          GenBank mRNA CJ059941.1 - 3 - 4 341 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001124.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          energy reserve metabolic process
          feeding behavior
          positive regulation of cAMP biosynthetic process
          protein localization to cell surface
          energy homeostasis
          corticotropin hormone receptor binding
          type 3 melanocortin receptor binding
          type 4 melanocortin receptor binding
          type 5 melanocortin receptor binding
          identical protein binding
          type 1 melanocortin receptor binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105140 NM_001177731.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105140 NM_001101482.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143296 NM_001177731.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143296 NM_001101482.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636372 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 557393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 590510 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469209 AK002812.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469209 AK004267.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469209 BC005501.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469209 NM_025422.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 929433 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887537 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851349 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168313 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 257221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470838 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 11269 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 217184 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 641842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 661925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151786 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 95251 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2143296 NM_001177731.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2143296 NM_001101482.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105140 NM_001177731.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105140 NM_001101482.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105135 NM_001177731.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105135 BQ175739.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105135 NM_001101482.2 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10587635 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587634 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587636 - -
          Affymetrix HT MG-430 PM Array Plate 1436274_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10587633 - -
          Mouse Genome 430 2.0 Array 1436274_PM_at - -
          Mouse Genome 430 2.0 Array 1436274_at - -
          Affymetrix Mouse Clariom S Array PSR0900008844.mm.1 NM_001101482.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008844.mm.1 NM_001101482.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008844.mm.1 NM_001177731.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008844.mm.1 NM_001177731.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008844.mm.1 TC0900001124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900008841.mm.1 NM_001177731.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008841.mm.1 NM_001177731.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008841.mm.1 TC0900001124.mm.1 MTA TC
          Mouse Expression Set 430 1436274_at - -
          Mouse Expression Set 430 1436274_PM_at - -
          Murine Genome U74v2 Set 131834_at - -
          Murine Genome U74v2 Set 131834_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436274_at - -
          Affymetrix HT Mouse Genome 430B Array 1436274_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0