Search
Search

Cluap1| Gene Symbol: Cluap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029738.2 | NP_084014.1 | 10 - 11 | 1160 | 59 | |
| GenBank mRNA | AK134648.1 | - | 10 - 11 | 1127 | 59 | |
| BC024910.1 | - | 10 - 11 | 1160 | 3603454 | 59 | |
| BC063769.1 | - | 4 - 5 | 394 | 59 | ||
Set Membership: |
|