Search
Search

Nfkbiz| Gene Symbol: Nfkbiz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159394.1 | NP_001152866.1 | 7 - 8 | 1704 | 67 | |
| NM_001159395.1 | NP_001152867.1 | 8 - 9 | 1586 | 67 | ||
| NM_030612.3 | NP_085115.1 | 9 - 10 | 1934 | 67 | ||
| GenBank mRNA | AB020974.1 | - | 9 - 10 | 2146 | 67 | |
| AB026551.2 | - | 7 - 8 | 1704 | 67 | ||
| AB047549.2 | - | 8 - 9 | 1599 | 67 | ||
| AK082908.1 | - | 7 - 8 | 1703 | 67 | ||
| AK136509.1 | - | 4 - 5 | 1173 | 67 | ||
| AK141318.1 | - | 7 - 8 | 1705 | 67 | ||
| AK149946.1 | - | 8 - 9 | 1582 | 67 | ||
| AK150519.1 | - | 7 - 8 | 1703 | 67 | ||
| AK152016.1 | - | 7 - 8 | 1705 | 67 | ||
| AK156826.1 | - | 7 - 8 | 1704 | 67 | ||
| AK159093.1 | - | 7 - 8 | 1705 | 67 | ||
| AK159171.1 | - | 7 - 8 | 1705 | 67 | ||
| AK159459.1 | - | 7 - 8 | 1705 | 67 | ||
| AK159645.1 | - | 7 - 8 | 1705 | 67 | ||
| AK159917.1 | - | 7 - 8 | 1705 | 67 | ||
| AK160065.1 | - | 7 - 8 | 1703 | 67 | ||
| BC058188.1 | - | 7 - 8 | 1689 | 5341736 | 67 | |
Set Membership: |
|