Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01171343_g1
          See other 2200002J24RIK GE Assays ›
          Gene Symbol:
          2200002J24Rik
          Gene Name:
          RIKEN cDNA 2200002J24 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 30698886 - 30710296 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01171343_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2200002J24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026961.2 NP_081237.1 3 - 4 324 75
          GenBank mRNA AK008620.1 - 3 - 4 295 75
          BC048568.1 - 3 - 4 324 6705641 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000571.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 2200002J24Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342629 BC048568.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342629 NM_026961.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342629 AK008620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342629 BC048568.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342629 NM_026961.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342629 AK008620.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472916 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96096 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134803 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342629 BC048568.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342629 NM_026961.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342629 AK008620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342629 BC048568.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342629 NM_026961.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342629 AK008620.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25032894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25032899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25032901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25032895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25032904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25032890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25032889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25032897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25032893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25032905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25032887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25032896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25032892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25032902 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134803 - -
          Whole Mouse Genome G4122A A_51_P342629 BC048568.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P342629 NM_026961.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P342629 AK008620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P342629 BC048568.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P342629 NM_026961.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P342629 AK008620.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134803 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06451742 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06480544 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06480548 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06451740 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06363026 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06480546 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05907620 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06363024 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06451737 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06451738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06480540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06366809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06363027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06480548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25032897 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552002 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552001 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552004 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552005 - -
          Affymetrix HT MG-430 PM Array Plate 1460478_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552000 - -
          Mouse Genome 430 2.0 Array 1460478_at - -
          Mouse Genome 430 2.0 Array 1460478_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69147_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69147_at - -
          Affymetrix Mouse Clariom S Array PSR0700005095.mm.1 NM_026961.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005095.mm.1 NM_026961.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005095.mm.1 TC0700000571.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005094.mm.1 NM_026961.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005094.mm.1 NM_026961.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005094.mm.1 TC0700000571.mm.1 MTA TC
          Mouse Expression Set 430 1460478_PM_at - -
          Mouse Expression Set 430 1460478_at - -
          Murine Genome U74v2 Set 166943_at - -
          Murine Genome U74v2 Set 166943_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460478_at - -
          Affymetrix HT Mouse Genome 430B Array 1460478_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 817548 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225349 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225349 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline