Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01171481_gH
          See other VMN2R83 GE Assays ›
          Gene Symbol:
          Vmn2r83
          Gene Name:
          vomeronasal 2, receptor 83
          Gene Aliases:
          EG625029
          Chromosome Location:
          Chr.10: 79468958 - 79492154 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01171481_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn2r83
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001104537.1 NP_001098007.1 2 - 3 506 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000805.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P615537 NM_001104537.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P615537 NM_001104537.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 62026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 110803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 695288 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634685 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104219 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826067 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526304 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 551287 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461543 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 903581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803077 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98887 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 146024 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68144 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45681 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P615537 NM_001104537.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P615537 NM_001104537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615537 NM_001104537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615537 NM_001104537.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27277272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27277230 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10364440 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364439 - -
          Affymetrix Mouse Gene 1.1 ST Array 10364438 - -
          Affymetrix Mouse Gene 1.1 ST Array 10364441 - -
          Affymetrix Mouse Clariom S Array PSR1000005825.mm.1 NM_001104537.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005825.mm.1 NM_001104537.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005825.mm.1 TC1000000805.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005823.mm.1 NM_001104537.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005823.mm.1 NM_001104537.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005823.mm.1 TC1000000805.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005824.mm.1 NM_001104537.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005824.mm.1 NM_001104537.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005824.mm.1 TC1000000805.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005826.mm.1 NM_001104537.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005826.mm.1 NM_001104537.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005826.mm.1 TC1000000805.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005821.mm.1 NM_001104537.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005821.mm.1 NM_001104537.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005821.mm.1 TC1000000805.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005822.mm.1 NM_001104537.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005822.mm.1 NM_001104537.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005822.mm.1 TC1000000805.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 392054 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline