Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01171664_g1
          See other DPPA5A GE Assays ›
          Gene Symbol:
          Dppa5a
          Gene Name:
          developmental pluripotency associated 5A
          Gene Aliases:
          AA536857, Dppa5, Esg1, ecat2
          Chromosome Location:
          Chr.9: 78367052 - 78368199 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01171664_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dppa5a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025274.3 NP_079550.2 1 - 2 219 66
          GenBank mRNA AF490349.1 - 1 - 2 175 66
          AK010519.1 - 1 - 2 176 66
          AK010584.1 - 1 - 2 175 66
          AK145761.1 - 1 - 2 178 66
          AV457404.1 - 1 - 2 221 66
          BC092353.1 - 1 - 2 164 30730511 66
          BC092354.1 - 1 - 2 172 30732471 66
          BC095979.1 - 1 - 2 164 30732937 66
          X57708.1 - 1 - 2 177 66

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dppa5a

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          regulation of gene expression
          nucleic acid binding
          RNA binding
          mRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028171 AK145761.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028171 AV476247.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028171 NM_025274.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028171 AF490349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028171 AK010584.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028171 AK145761.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028171 AV476247.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028171 NM_025274.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028171 AF490349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028171 AK010584.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P18706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26658897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26658886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26658880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26658879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26658884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26658881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26658895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26658894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26658898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26658870 - -
          Whole Mouse Genome G4122A A_52_P18706 - -
          Whole Mouse Genome G4122A A_52_P362403 AK145761.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362403 AV476247.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362403 NM_025274.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362403 AF490349.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362403 AK010519.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120179 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07864806 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08118729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08100997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07679758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08031987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08100998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08118729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08070478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08102979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08070474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659195 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10520418 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595157 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595158 - -
          Affymetrix HT MG-430 PM Array Plate 1416552_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10595156 - -
          Affymetrix Mouse Gene 1.1 ST Array 10520417 - -
          Mouse Genome 430 2.0 Array 1416552_PM_at - -
          Mouse Genome 430 2.0 Array 1416552_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 434423_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 434423_at - -
          Affymetrix Mouse Clariom S Array PSR0900022451.mm.1 NM_025274.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022451.mm.1 NM_025274.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022451.mm.1 TC0900002720.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900022455.mm.1 NM_025274.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022455.mm.1 NM_025274.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022455.mm.1 TC0900002720.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900022453.mm.1 NM_025274.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022453.mm.1 NM_025274.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022453.mm.1 TC0900002720.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900022452.mm.1 NM_025274.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022452.mm.1 NM_025274.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022452.mm.1 TC0900002720.mm.1 MTA TC
          Mouse Expression Set 430 1416552_PM_at - -
          Mouse Expression Set 430 1416552_at - -
          Mouse Genome 430A 2.0 Array 1416552_PM_at - -
          Mouse Genome 430A 2.0 Array 1416552_at - -
          Affymetrix HT Mouse Genome 430A Array 1416552_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416552_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 896585 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1580450 - -
          Illumina mouse-6 v1.0 expression beadchip 1580450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001580450 - -
          mouse-6 v1.1 (Illumina) scl013915.2_46-S - -
          Illumina Sentrix mouse-6.v1 bead array 1580450 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl013915.2_46-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1580450 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2774896 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3018711 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3090545 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3018711 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3090545 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2774896 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline