Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01171709_s1
          See other VMN1R179 GE Assays ›
          Gene Symbol:
          Vmn1r179
          Gene Name:
          vomeronasal 1 receptor 179
          Gene Aliases:
          V1rd17
          Chromosome Location:
          Chr.7: 23928386 - 23929357 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01171709_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r179
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207545.1 NP_997428.1 1 859 145

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r179

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2108521 NM_207545.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2108521 NM_207545.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002938 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06139897 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107537 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06139897 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06406676 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06363410 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06363419 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06406682 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06363421 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06406693 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06363412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06363401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06406695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06406676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06603803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06360273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05909528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002929 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550823 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550822 - -
          Affymetrix Mouse Clariom S Array PSR0700002629.mm.1 NM_207545.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002629.mm.1 NM_207545.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002629.mm.1 TC0700000361.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 824407 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4060161 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004060161 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2417982 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2417982 - -
          mouse-6 v1.1 (Illumina) scl0404286.1_268-S - -
          Illumina Sentrix mouse-6.v1 bead array 4060161 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0404286.1_268-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4060161 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2417982 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2417982 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline