Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01171976_s1
          See other OLFR867 GE Assays ›
          Gene Symbol:
          Olfr867
          Gene Name:
          olfactory receptor 867
          Gene Aliases:
          MOR143-2
          Chromosome Location:
          Chr.9: 20054466 - 20055461 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01171976_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr867
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011748.1 NP_001011748.1 1 724 149

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr867

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P433753 NM_001011748.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P433753 NM_001011748.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P433753 NM_001011748.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P433753 NM_001011748.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P433753 NM_001011748.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P433753 NM_001011748.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341628 - -
          Whole Mouse Genome G4122A A_51_P433753 NM_001011748.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433753 NM_001011748.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06715693 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07921207 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07794149 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07794147 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07794151 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07921204 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07794144 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07500964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07921207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07640471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07921206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07567258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341635 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10591215 - -
          Affymetrix Mouse Gene 1.1 ST Array 10591214 - -
          Affymetrix Mouse Clariom S Array PSR0900015247.mm.1 NM_001011748.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015247.mm.1 NM_001011748.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015247.mm.1 TC0900001828.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 498736 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161367 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161367 - -
          mouse-6 v1.1 (Illumina) GI_58801283-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161367 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161367 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline