Search
Search

Zfp202| Gene Symbol: Zfp202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030713.2 | NP_109638.2 | 2 - 3 | 614 | 69 | |
| GenBank mRNA | AF292648.1 | - | 2 - 3 | 604 | 69 | |
| AK170181.1 | - | 2 - 3 | 604 | 69 | ||
| BC049772.1 | - | 2 - 3 | 542 | 6334069 | 69 | |
Set Membership: |
|