Search
Search
| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB093224.1 | - | 6 - 7 | 2197 | 69 | |
| AF363690.1 | - | 11 - 12 | 2927 | 69 | ||
| AK004685.1 | - | 2 - 3 | 326 | 69 | ||
| AK082152.1 | - | 1 - 2 | 89 | 69 | ||
| AK147276.1 | - | 11 - 12 | 3560 | 69 | ||
| AK147535.1 | - | 12 - 13 | 5678 | 69 | ||
| BC040463.1 | - | 2 - 3 | 627 | 69 | ||
| BC058665.1 | - | 8 - 9 | 2445 | 69 | ||
Set Membership: |
|