Search
Search
| Gene Symbol: Cd3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009850.2 | NP_033980.1 | 5 - 6 | 599 | 75 | |
| GenBank mRNA | AK153980.1 | - | 5 - 6 | 531 | 75 | |
| AK153987.1 | - | 5 - 6 | 518 | 75 | ||
| BC027528.1 | - | 5 - 6 | 530 | 1494674 | 75 | |
| EF474453.1 | - | 5 - 6 | 488 | 75 | ||
| GU456630.1 | - | 5 - 6 | 488 | 75 | ||
| M58149.1 | - | 5 - 6 | 505 | 75 | ||
| Y00635.1 | - | 5 - 6 | 627 | 75 | ||
Set Membership: |
|