Search
Search

Tex12| Gene Symbol: Tex12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025687.3 | NP_079963.1 | 3 - 4 | 218 | 130 | |
| GenBank mRNA | AF285582.1 | - | 3 - 4 | 276 | 130 | |
| AK006285.1 | - | 3 - 4 | 262 | 130 | ||
| AK014896.1 | - | 3 - 4 | 275 | 130 | ||
| AK017776.1 | - | 3 - 4 | 274 | 130 | ||
| BC048458.1 | - | 3 - 4 | 218 | 6775116 | 130 | |
| BC061081.1 | - | 3 - 4 | 273 | 6773183 | 130 | |
Set Membership: |
|