| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900006394.mm.1 |
NM_016688.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006394.mm.1 |
NM_016688.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006394.mm.1 |
TC0900000839.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006396.mm.1 |
NM_016688.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006396.mm.1 |
NM_016688.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006396.mm.1 |
TC0900000839.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006395.mm.1 |
NM_016688.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006395.mm.1 |
NM_016688.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006395.mm.1 |
TC0900000839.mm.1 |
MTA TC |